Assay Details
Gene Symbol: Plat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013151.2 | NP_037283.2 | 2 - 3 | 174 | 63 | |
XM_006253347.2 | XP_006253409.1 | 2 - 3 | 246 | 63 | ||
XM_008771348.1 | XP_008769570.1 | 3 - 4 | 304 | 63 | ||
GenBank mRNA | BC061565.1 | - | 2 - 3 | 174 | 63 | |
M23697.1 | - | 1 - 2 | 98 | 63 |
Gene Symbol: LOC100910418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751623.3 | XP_003751671.2 | 2 - 3 | 178 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
