Assay Details
Gene Symbol: Gtf2e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253230.2 | XP_006253292.1 | 1 - 2 | 102 | 68 | |
XM_006253231.2 | XP_006253293.1 | 1 - 2 | 102 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006253230.2, XM_006253231.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
