Assay Details
Gene Symbol: Gtf2e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107318.2 | NP_001100788.2 | 7 - 8 | 798 | 106 | |
XM_006253230.2 | XP_006253292.1 | 7 - 8 | 863 | 106 | ||
GenBank mRNA | CF110694.1 | - | 7 - 8 | 814 | 106 | |
FQ225698.1 | - | 6 - 7 | 774 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1028065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105024 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1028065 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1028065 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1028065 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P290767 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1028065 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94352 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10792059 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792056 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792057 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792055 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792058 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792053 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792052 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792054 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10792051 | - | - |
Rat Genome 230 2.0 Array | 1399114_at | - | - |
Rat Genome 230 2.0 Array | 1399114_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1399114_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1399114_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1600013199.rn.1 | TC1600001795.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1600013203.rn.1 | TC1600001795.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1600013200.rn.1 | TC1600001795.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1600013198.rn.1 | TC1600001795.rn.1 | RTA TC |
Rat Expression Set 230 | 1399114_PM_at | - | - |
Rat Expression Set 230 | 1399114_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10792051 | - | - |
Rat Genome U34 Set | rc_AA996974_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20942786 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375017 | - | - |
