Assay Details
Gene Symbol: Ankrd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253458.2 | XP_006253520.1 | 3 - 4 | 264 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006253458.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
