Assay Details
Gene Symbol: Ankrd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253453.2 | XP_006253515.1 | 3 - 4 | 859 | 79 | |
XM_006253454.2 | XP_006253516.1 | 3 - 4 | 1200 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
