Assay Details
Gene Symbol: Tnfsf13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253480.2 | XP_006253542.1 | 5 - 6 | 1342 | 106 | |
XM_006253481.2 | XP_006253543.1 | 4 - 5 | 1285 | 106 | ||
XM_006253482.2 | XP_006253544.1 | 4 - 5 | 1245 | 106 | ||
XM_006253483.2 | XP_006253545.1 | 3 - 4 | 1187 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
