Assay Details
Gene Symbol: Tnfsf13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253480.2 | XP_006253542.1 | 6 - 7 | 1496 | 62 | |
XM_006253481.2 | XP_006253543.1 | 5 - 6 | 1439 | 62 | ||
XM_006253482.2 | XP_006253544.1 | 5 - 6 | 1399 | 62 | ||
XM_006253483.2 | XP_006253545.1 | 4 - 5 | 1341 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
