Assay Details
Gene Symbol: Smim19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728416.4 | XP_002728462.1 | 2 - 3 | 391 | 106 | |
GenBank mRNA | FQ214349.1 | - | 2 - 3 | 362 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
