Assay Details
Gene Symbol: Agpat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253381.2 | XP_006253443.1 | 1 - 2 | 480 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
