Assay Details
Gene Symbol: Uck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102408.1 | NP_001095878.1 | 1 - 2 | 547 | 81 | |
GenBank mRNA | BC167060.1 | - | 1 - 2 | 298 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
