Assay Details
Gene Symbol: Uck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102408.1 | NP_001095878.1 | 4 - 5 | 938 | 125 | |
GenBank mRNA | AB030700.1 | - | 1 - 2 | 40 | 125 | |
BC167060.1 | - | 4 - 5 | 689 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P14144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5428 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14144 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14144 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P14144 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P503428 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P14144 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20469 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 61004 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10769623 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769626 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769627 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769625 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769622 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769624 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10769621 | - | - |
Rat Genome 230 2.0 Array | 1383945_at | - | - |
Rat Genome 230 2.0 Array | 1383945_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383945_at | - | - |
Affymetrix HT Rat Focus Array | 1383945_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300013113.rn.1 | TC1300001854.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300013110.rn.1 | TC1300001854.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300013117.rn.1 | TC1300001854.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300013111.rn.1 | TC1300001854.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300013115.rn.1 | TC1300001854.rn.1 | RTA TC |
Rat Expression Set 230 | 1383945_PM_at | - | - |
Rat Expression Set 230 | 1383945_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769621 | - | - |
Rat Genome U34 Set | rc_AA859827_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20780474 | - | - |
