Assay Details
Gene Symbol: Glul | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017073.3 | NP_058769.4 | 4 - 5 | 592 | 82 | |
GenBank mRNA | BC061559.1 | - | 4 - 5 | 605 | 82 | |
BC072694.1 | - | 4 - 5 | 592 | 82 | ||
FQ213256.1 | - | 4 - 5 | 603 | 82 | ||
FQ216093.1 | - | 4 - 5 | 597 | 82 | ||
FQ226911.1 | - | 4 - 5 | 615 | 82 | ||
FQ227474.1 | - | 4 - 5 | 604 | 82 | ||
FQ232853.1 | - | 4 - 5 | 653 | 82 | ||
FQ232963.1 | - | 4 - 5 | 604 | 82 | ||
FQ233770.1 | - | 4 - 5 | 604 | 82 | ||
FQ234289.1 | - | 4 - 5 | 605 | 82 | ||
M29579.1 | - | 4 - 5 | 603 | 82 | ||
M91652.1 | - | 4 - 5 | 601 | 82 | ||
X07921.1 | - | 4 - 5 | 603 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
