Assay Details
Gene Symbol: Ccnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107171.1 | NP_001100641.1 | 2 - 3 | 269 | 113 | |
XM_006249686.2 | XP_006249748.1 | 2 - 3 | 277 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
