Assay Details
Gene Symbol: Ccnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107171.1 | NP_001100641.1 | 7 - 8 | 741 | 106 | |
XM_006249686.2 | XP_006249748.1 | 7 - 8 | 749 | 106 | ||
XM_006249687.2 | XP_006249749.1 | 3 - 4 | 3450 | 106 | ||
XM_008769457.1 | XP_008767679.1 | 6 - 7 | 620 | 106 | ||
XM_008769458.1 | XP_008767680.1 | 4 - 5 | 2526 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
