Assay Details
Gene Symbol: Cd247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170789.2 | NP_740770.2 | 3 - 4 | 340 | 80 | |
XM_006250147.2 | XP_006250209.1 | 3 - 4 | 383 | 80 | ||
GenBank mRNA | BC097933.1 | - | 3 - 4 | 283 | 80 | |
L08447.1 | - | 3 - 4 | 267 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
