Assay Details
Gene Symbol: Ildr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769713.1 | XP_008767935.1 | 4 - 5 | 728 | 62 | |
XM_008769714.1 | XP_008767936.1 | 4 - 5 | 667 | 62 | ||
XM_008769716.1 | XP_008767938.1 | 4 - 5 | 2662 | 62 | ||
XM_008769717.1 | XP_008767939.1 | 4 - 5 | 2640 | 62 | ||
XM_008769718.1 | XP_008767940.1 | 4 - 5 | 532 | 62 | ||
XM_008769719.1 | XP_008767941.1 | 4 - 5 | 728 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
