Assay Details
Gene Symbol: Ildr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769713.1 | XP_008767935.1 | 7 - 8 | 1170 | 138 | |
XM_008769714.1 | XP_008767936.1 | 7 - 8 | 1109 | 138 | ||
XM_008769715.1 | XP_008767937.1 | 6 - 7 | 1113 | 138 | ||
XM_008769716.1 | XP_008767938.1 | 7 - 8 | 3104 | 138 | ||
XM_008769717.1 | XP_008767939.1 | 7 - 8 | 3082 | 138 | ||
XM_008769718.1 | XP_008767940.1 | 7 - 8 | 974 | 138 | ||
XM_008769719.1 | XP_008767941.1 | 6 - 7 | 993 | 138 | ||
XM_008769720.1 | XP_008767942.1 | 4 - 5 | 789 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
