Assay Details
Gene Symbol: Rcsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250181.2 | XP_006250243.1 | 2 - 3 | 319 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
