Assay Details
Gene Symbol: Rxrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031765.1 | NP_113953.1 | 3 - 4 | 673 | 75 | |
XM_006250212.2 | XP_006250274.1 | 2 - 3 | 817 | 75 | ||
GenBank mRNA | AJ223083.1 | - | 2 - 3 | 208 | 75 | |
BC091363.1 | - | 3 - 4 | 673 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
