Assay Details
Gene Symbol: Rxrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031765.1 | NP_113953.1 | 5 - 6 | 1010 | 62 | |
XM_006250212.2 | XP_006250274.1 | 4 - 5 | 1154 | 62 | ||
GenBank mRNA | AF059312.1 | - | 1 - 2 | 138 | 62 | |
AJ223083.1 | - | 4 - 5 | 545 | 62 | ||
BC091363.1 | - | 5 - 6 | 1010 | 62 |
Gene Symbol: LOC102553986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250213.2 | XP_006250275.1 | 1 - 2 | 1615 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
