Assay Details
Gene Symbol: Rgsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769723.1 | XP_008767945.1 | 2 - 3 | 391 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
