Assay Details
Gene Symbol: Adipor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207587.1 | NP_997470.1 | 2 - 3 | 363 | 125 | |
XM_006249852.2 | XP_006249914.1 | 2 - 3 | 399 | 125 | ||
XM_008769543.1 | XP_008767765.1 | 2 - 3 | 700 | 125 | ||
XM_008769544.1 | XP_008767766.1 | 2 - 3 | 1062 | 125 | ||
GenBank mRNA | BC061838.1 | - | 2 - 3 | 363 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
