Assay Details
Gene Symbol: Adipor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207587.1 | NP_997470.1 | 3 - 4 | 481 | 65 | |
XM_006249852.2 | XP_006249914.1 | 3 - 4 | 517 | 65 | ||
XM_008769543.1 | XP_008767765.1 | 3 - 4 | 818 | 65 | ||
XM_008769544.1 | XP_008767766.1 | 3 - 4 | 1180 | 65 | ||
GenBank mRNA | BC061838.1 | - | 3 - 4 | 481 | 5599938 | 65 |
DQ148391.1 | - | 2 - 3 | 269 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
