Assay Details
Gene Symbol: Mpzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007728.1 | NP_001007729.1 | 2 - 3 | 358 | 73 | |
XM_006250180.2 | XP_006250242.1 | 2 - 3 | 406 | 73 | ||
GenBank mRNA | BC078916.1 | - | 2 - 3 | 358 | 7096438 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
