Assay Details
Gene Symbol: Mpzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007728.1 | NP_001007729.1 | 3 - 4 | 571 | 118 | |
XM_006250180.2 | XP_006250242.1 | 3 - 4 | 619 | 118 | ||
GenBank mRNA | BC078916.1 | - | 3 - 4 | 571 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
