Assay Details
Gene Symbol: Mpzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007728.1 | NP_001007729.1 | 5 - 6 | 812 | 59 | |
GenBank mRNA | BC078916.1 | - | 5 - 6 | 812 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P521699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122002 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P521699 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P521699 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P521699 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P552926 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P521699 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50603 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47488 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10769541 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769542 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769540 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769544 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769539 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769543 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10769538 | - | - |
Rat Genome 230 2.0 Array | 1390114_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390114_at | - | - |
Affymetrix HT Rat Focus Array | 1390114_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390114_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300012950.rn.1 | TC1300001831.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300012949.rn.1 | TC1300001831.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300012951.rn.1 | TC1300001831.rn.1 | RTA TC |
Rat Expression Set 230 | 1390114_PM_at | - | - |
Rat Expression Set 230 | 1390114_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10769538 | - | - |
Rat Genome U34 Set | rc_AA875232_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21768690 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372608 | - | - |
RatRef-12 Expression BeadChip | ILMN_66207 | - | - |
