Assay Details
Gene Symbol: Ptgs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017232.3 | NP_058928.3 | 2 - 3 | 162 | 124 | |
GenBank mRNA | L20085.1 | - | 2 - 3 | 162 | 124 | |
L25925.1 | - | 2 - 3 | 168 | 124 | ||
U04300.1 | - | 2 - 3 | 162 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
