Assay Details
Gene Symbol: Pogk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250161.2 | XP_006250223.1 | 1 - 2 | 142 | 91 | |
XM_006250162.2 | XP_006250224.1 | 1 - 2 | 142 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006250161.2, XM_006250162.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
