Assay Details
Gene Symbol: Pogk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107194.2 | NP_001100664.2 | 2 - 3 | 232 | 102 | |
XM_006250161.2 | XP_006250223.1 | 3 - 4 | 606 | 102 | ||
XM_006250162.2 | XP_006250224.1 | 3 - 4 | 471 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
