Assay Details
Gene Symbol: Gorab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100563.1 | NP_001094033.1 | 1 - 2 | 145 | 86 | |
GenBank mRNA | BC160825.1 | - | 1 - 2 | 145 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
