Assay Details
Gene Symbol: Gorab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100563.1 | NP_001094033.1 | 3 - 4 | 595 | 130 | |
XM_008769650.1 | XP_008767872.1 | 2 - 3 | 1171 | 130 | ||
GenBank mRNA | BC086370.1 | - | 1 - 2 | 74 | 130 | |
BC160825.1 | - | 3 - 4 | 595 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
