Assay Details
Gene Symbol: Gorab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100563.1 | NP_001094033.1 | 4 - 5 | 740 | 87 | |
XM_008769650.1 | XP_008767872.1 | 3 - 4 | 1316 | 87 | ||
GenBank mRNA | BC086370.1 | - | 2 - 3 | 219 | 87 | |
BC160825.1 | - | 4 - 5 | 740 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
