Assay Details
Gene Symbol: Cyb5r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013126.1 | NP_001013144.1 | 8 - 9 | 790 | 69 | |
GenBank mRNA | BC089945.1 | - | 8 - 9 | 790 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
