Assay Details
Gene Symbol: Il10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012854.2 | NP_036986.2 | 3 - 4 | 382 | 73 | |
XM_006249712.2 | XP_006249774.1 | 3 - 4 | 721 | 73 | ||
GenBank mRNA | L02926.1 | - | 3 - 4 | 382 | 73 | |
X60675.1 | - | 425 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
