Assay Details
Gene Symbol: Pak6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106498.1 | NP_001099968.1 | 3 - 4 | 636 | 67 | |
XM_008762091.1 | XP_008760313.1 | 2 - 3 | 1936 | 67 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001106498.1, XM_008762091.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
