Assay Details
Gene Symbol: Pak6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106498.1 | NP_001099968.1 | 4 - 5 | 845 | 74 | |
XM_008762091.1 | XP_008760313.1 | 3 - 4 | 2145 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
