Assay Details
Gene Symbol: Pak6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106498.1 | NP_001099968.1 | 7 - 8 | 2125 | 66 | |
XM_008762091.1 | XP_008760313.1 | 6 - 7 | 3425 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
