Assay Details
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031793.1 | NP_113981.1 | 5 - 6 | 466 | 129 | |
XM_006234324.2 | XP_006234386.1 | 5 - 6 | 842 | 129 | ||
XM_006234325.2 | XP_006234387.1 | 5 - 6 | 663 | 129 | ||
XM_006234326.2 | XP_006234388.1 | 5 - 6 | 786 | 129 | ||
XM_006234327.2 | XP_006234389.1 | 4 - 5 | 738 | 129 | ||
XM_006234328.2 | XP_006234390.1 | 5 - 6 | 733 | 129 | ||
XM_006234330.2 | XP_006234392.1 | 4 - 5 | 559 | 129 | ||
XM_006234331.2 | XP_006234393.1 | 4 - 5 | 685 | 129 | ||
XM_006234333.2 | XP_006234395.1 | 4 - 5 | 699 | 129 | ||
XM_006234334.2 | XP_006234396.1 | 4 - 5 | 520 | 129 | ||
XM_006234335.2 | XP_006234397.1 | 4 - 5 | 646 | 129 | ||
XM_008761930.1 | XP_008760152.1 | 4 - 5 | 762 | 129 | ||
XM_008761931.1 | XP_008760153.1 | 4 - 5 | 723 | 129 | ||
GenBank mRNA | AF043642.1 | - | 5 - 6 | 466 | 129 | |
BC085723.1 | - | 4 - 5 | 512 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
