Assay Details
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031793.1 | NP_113981.1 | 7 - 8 | 622 | 104 | |
XM_006234324.2 | XP_006234386.1 | 7 - 8 | 998 | 104 | ||
XM_006234325.2 | XP_006234387.1 | 7 - 8 | 819 | 104 | ||
XM_006234326.2 | XP_006234388.1 | 7 - 8 | 942 | 104 | ||
XM_006234327.2 | XP_006234389.1 | 6 - 7 | 894 | 104 | ||
XM_006234328.2 | XP_006234390.1 | 7 - 8 | 889 | 104 | ||
XM_006234330.2 | XP_006234392.1 | 6 - 7 | 715 | 104 | ||
XM_006234331.2 | XP_006234393.1 | 6 - 7 | 841 | 104 | ||
XM_006234333.2 | XP_006234395.1 | 6 - 7 | 855 | 104 | ||
XM_006234334.2 | XP_006234396.1 | 6 - 7 | 676 | 104 | ||
XM_006234335.2 | XP_006234397.1 | 6 - 7 | 802 | 104 | ||
XM_008761930.1 | XP_008760152.1 | 6 - 7 | 918 | 104 | ||
XM_008761931.1 | XP_008760153.1 | 6 - 7 | 879 | 104 | ||
GenBank mRNA | AF043642.1 | - | 7 - 8 | 622 | 104 | |
BC085723.1 | - | 6 - 7 | 668 | 104 | ||
FQ225838.1 | - | 2 - 3 | 137 | 104 | ||
FQ230786.1 | - | 2 - 3 | 137 | 104 | ||
FQ232477.1 | - | 2 - 3 | 137 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
