Assay Details
Gene Symbol: Ndufs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106489.1 | NP_001099959.1 | 1 - 2 | 246 | 77 | |
GenBank mRNA | FQ212516.1 | - | 1 - 2 | 70 | 77 | |
FQ219879.1 | - | 1 - 2 | 73 | 77 | ||
FQ229748.1 | - | 1 - 2 | 93 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
