Assay Details
Gene Symbol: Nup35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004229.1 | NP_001004229.1 | 2 - 3 | 245 | 127 | |
XM_006234430.2 | XP_006234492.1 | 2 - 3 | 252 | 127 | ||
GenBank mRNA | BC078914.1 | - | 2 - 3 | 245 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
