Assay Details
Gene Symbol: Arhgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107747.2 | NP_001101217.2 | 11 - 12 | 1159 | 72 | |
XM_006234551.1 | XP_006234613.1 | 12 - 13 | 1196 | 72 | ||
XM_006234552.2 | XP_006234614.1 | 12 - 13 | 1226 | 72 | ||
XM_006234553.2 | XP_006234615.1 | 12 - 13 | 1196 | 72 | ||
XM_006234554.1 | XP_006234616.1 | 13 - 14 | 1238 | 72 | ||
XM_006234556.1 | XP_006234618.1 | 12 - 13 | 1173 | 72 | ||
XM_006234557.2 | XP_006234619.1 | 12 - 13 | 1203 | 72 | ||
XM_006234558.2 | XP_006234620.1 | 11 - 12 | 1144 | 72 | ||
XM_006234559.2 | XP_006234621.1 | 9 - 10 | 2582 | 72 | ||
XM_006234560.2 | XP_006234622.1 | 11 - 12 | 1185 | 72 | ||
GenBank mRNA | DN932805.1 | - | 3 - 4 | 319 | 72 | |
EV768858.1 | - | 7 - 8 | 630 | 72 | ||
FQ224002.1 | - | 11 - 12 | 1119 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
