Assay Details
Gene Symbol: Arhgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107747.2 | NP_001101217.2 | 7 - 8 | 768 | 74 | |
XM_006234551.1 | XP_006234613.1 | 8 - 9 | 805 | 74 | ||
XM_006234552.2 | XP_006234614.1 | 8 - 9 | 835 | 74 | ||
XM_006234553.2 | XP_006234615.1 | 8 - 9 | 805 | 74 | ||
XM_006234554.1 | XP_006234616.1 | 9 - 10 | 847 | 74 | ||
XM_006234556.1 | XP_006234618.1 | 8 - 9 | 782 | 74 | ||
XM_006234557.2 | XP_006234619.1 | 8 - 9 | 812 | 74 | ||
XM_006234558.2 | XP_006234620.1 | 7 - 8 | 753 | 74 | ||
XM_006234559.2 | XP_006234621.1 | 5 - 6 | 2191 | 74 | ||
XM_006234560.2 | XP_006234622.1 | 7 - 8 | 794 | 74 | ||
GenBank mRNA | EV768858.1 | - | 3 - 4 | 239 | 74 | |
FQ224002.1 | - | 7 - 8 | 728 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
