Assay Details
Gene Symbol: Arhgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107747.2 | NP_001101217.2 | 1 - 2 | 115 | 60 | |
GenBank mRNA | CK480577.1 | - | 1 - 2 | 115 | 60 | |
FQ224002.1 | - | 1 - 2 | 75 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107747.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
