Assay Details
Gene Symbol: Hsd17b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032066.1 | NP_114455.1 | 7 - 8 | 551 | 59 | |
XM_006234599.2 | XP_006234661.1 | 7 - 8 | 718 | 59 | ||
GenBank mRNA | BC061543.1 | - | 7 - 8 | 572 | 6888949 | 59 |
FQ213557.1 | - | 7 - 8 | 587 | 59 | ||
FQ214777.1 | - | 7 - 8 | 616 | 59 | ||
U81186.1 | - | 7 - 8 | 551 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
