Assay Details
Gene Symbol: Mavs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005556.1 | NP_001005556.1 | 2 - 3 | 406 | 73 | |
XM_006235034.2 | XP_006235096.1 | 2 - 3 | 274 | 73 | ||
XM_006235035.2 | XP_006235097.1 | 2 - 3 | 988 | 73 | ||
XM_006235036.2 | XP_006235098.1 | 3 - 4 | 307 | 73 | ||
XM_008762179.1 | XP_008760401.1 | 2 - 3 | 1481 | 73 | ||
XM_008762180.1 | XP_008760402.1 | 2 - 3 | 294 | 73 | ||
GenBank mRNA | BC081869.1 | - | 2 - 3 | 406 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
