Assay Details
Gene Symbol: Mavs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005556.1 | NP_001005556.1 | 4 - 5 | 750 | 83 | |
XM_006235034.2 | XP_006235096.1 | 4 - 5 | 618 | 83 | ||
XM_006235035.2 | XP_006235097.1 | 4 - 5 | 1332 | 83 | ||
XM_006235036.2 | XP_006235098.1 | 5 - 6 | 651 | 83 | ||
XM_006235037.2 | XP_006235099.1 | 3 - 4 | 654 | 83 | ||
XM_008762179.1 | XP_008760401.1 | 4 - 5 | 1825 | 83 | ||
XM_008762180.1 | XP_008760402.1 | 4 - 5 | 638 | 83 | ||
GenBank mRNA | BC081869.1 | - | 4 - 5 | 750 | 83 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_006235037.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
