Assay Details
Gene Symbol: Bahd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234794.2 | XP_006234856.1 | 2 - 3 | 1923 | 77 | |
XM_006234796.2 | XP_006234858.1 | 2 - 3 | 3724 | 77 | ||
XM_008762115.1 | XP_008760337.1 | 2 - 3 | 1593 | 77 | ||
XM_008762117.1 | XP_008760339.1 | 2 - 3 | 1593 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
