Assay Details
Gene Symbol: Bahd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234794.2 | XP_006234856.1 | 4 - 5 | 2465 | 62 | |
XM_006234796.2 | XP_006234858.1 | 4 - 5 | 4266 | 62 | ||
XM_008762115.1 | XP_008760337.1 | 4 - 5 | 2135 | 62 | ||
XM_008762116.1 | XP_008760338.1 | 4 - 5 | 2132 | 62 | ||
XM_008762117.1 | XP_008760339.1 | 4 - 5 | 2135 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
