Assay Details
Gene Symbol: Bahd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234794.2 | XP_006234856.1 | 5 - 6 | 2539 | 90 | |
XM_006234796.2 | XP_006234858.1 | 5 - 6 | 4340 | 90 | ||
XM_008762115.1 | XP_008760337.1 | 5 - 6 | 2209 | 90 | ||
XM_008762116.1 | XP_008760338.1 | 5 - 6 | 2206 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
